Bromoacetonitrile ≥96%

Supplier: BeanTown Chemical

121745-10G 121745-250G 121745-50G
BT121745-10GEA 37.55 USD
BT121745-10G BT121745-250G BT121745-50G
Bromoacetonitrile ≥96%
Bromoacetonitrile

CAS: 590-17-0; EC No: 209-672-1; MDL No: MFCD00001884; RTECS: AL7970000 UN No: UN2922; Haz Class: 8 (6.1); Packing Group: II Liquid; Linear Formula: BrCH2CN; MW: 119.95 Boiling Point: 560-562°/24 mmHg; Flash point: 81°C (178°F) Density (g/mL): 1.722; Refractive Index: 1.479


  • RTECS: AL7970000
  • Refractive index: 1.479
  • Liquid

Formula: BrCNCH₂
MW: 119.95 g/mol
Boiling Pt: 150
Melting Pt: -36
Density: 1.722 g/mL
Flash Pt: 81 °C (178 °F)
MDL Number: MFCD00001884
CAS Number: 590-17-0
EINECS: 209-672-1
UN: 2922
ADR: 6.1 II

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR