Iron ≥98% (trace metals basis), spherical powder 1-3 µm

Supplier: BeanTown Chemical

121470-100G 121470-25G
BT121470-100GEA 95 USD
BT121470-100G BT121470-25G
Iron ≥98% (trace metals basis), spherical powder 1-3 µm
Iron

CAS: 7439-89-6; EC No: 231-096-4; MDL No: MFCD00010999; RTECS: NO4565500 UN No: UN3089; Haz Class: 4.1; Packing Group: II Powder; Molecular Formula: Fe; MW: 55.85 Melting Point: 1535°; Boiling Point: 2750° Density (g/mL): 7.86


  • RTECS: NO4565500
  • Powder

Formula: Fe
MW: 55.85 g/mol
Boiling Pt: 3000
Melting Pt: 1535
Density: 7.86 g/mL
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
UN: 3089
ADR: 4.1 II
Merck Index: 13,05109

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR