Bis(cyclopentadienyl)zirconium dichloride
Supplier: BeanTown Chemical
Synonyms:
Zirconocene dichloride
121075-100G
121075-25G
121075-5G
BT121075-100GEA
333
USD
BT121075-100G
BT121075-25G
BT121075-5G
Bis(cyclopentadienyl)zirconium dichloride
Bis(cyclopentadienyl)zirconium dichloride
CAS: 1291-32-3; EC No: 215-066-8; MDL No: MFCD00003726
UN No: UN3096; Haz Class: 8 (4.3); Packing Group: II
Powder; Molecular Formula: C10H10Cl2Zr ; MW: 292.32
Melting Point: 242-245°
Air Sensitive, Moisture Sensitive
- Air Sensitive, Moisture Sensitive
- Powder
Formula:
C₁₀H₁₀Cl₂Zr Melting Pt: 242…245 °C |
MDL Number:
MFCD00003726 CAS Number: 1291-32-3 EINECS: 215-066-8 UN: 3096 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...