Valacyclovir hydrochloride hydrate ≥97.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
L-Valine 2-(2-Amino-1,6-dihydro-6-oxo-9H-purin-9-ylmethoxy)ethyl Ester Hydrochloride
CAS Number: 124832-27-5
MDL Number: MFCD01861507
Molecular Formula: C13H20N6O4
Molecular Weight: 360.80
Purity/Analysis Method: >97.0% (HPLC,N)
Form: Crystal
Color: White
Lambda max.: 253 nm (H2O)
Product Categories: Bioscience, Antiviral Ingredients, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleosides and Their Analogs, Prodrug Ingredients
Other Product Names / Synonyms: L-Valine 2-(2-Amino-1,6-dihydro-6-oxo-9H-purin-9-ylmethoxy)ethyl Ester Hydrochloride
Formula:
C₁₃H₂₀N₆O₄ MW: 360.80 g/mol |
MDL Number:
MFCD01861507 CAS Number: 124832-27-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...