2-(3-Trifluoromethylphenyl)ethylamine ≥96.0%

Supplier: TCI America

Synonyms: 3-(Trifluoromethyl)phenethylamine

T3086-1G T3086-5G
TCT3086-1GEA 48 USD
TCT3086-1G TCT3086-5G
2-(3-Trifluoromethylphenyl)ethylamine ≥96.0%
2-(3-Trifluoromethylphenyl)ethylamine

CAS Number: 52516-30-0 MDL Number: MFCD00040756 Molecular Formula: C9H10F3N Molecular Weight: 189.18 Purity/Analysis Method: 96.0% (GC,T) Form: Clear Liquid Color: Slightly Pale Yellow Specific Gravity (20/20): 1.20


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks Other Product Names / Synonyms: 3-(Trifluoromethyl)phenethylamine

Formula: C₉H₁₀F₃N
MW: 189.18 g/mol
Density: 1.20 (20 °C)
MDL Number: MFCD00040756
CAS Number: 52516-30-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f24d448672eabaebde309db5abb1e847baa02fb4&vid=f24d448672eabaebde309db5abb1e847baa02fb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR