3-(3-Trifluoromethylphenyl)propionic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
3-(3-(Trifluoromethyl)phenyl)propanoic acid, 3-[3-(Trifluoromethyl)phenyl]propionic acid, 3-(Trifluoromethyl)hydrocinnamic acid
T2931-1G
T2931-5G
TCT2931-1GEA
22
USD
TCT2931-1G
TCT2931-5G
3-(3-Trifluoromethylphenyl)propionic acid ≥98.0% (by GC, titration analysis)
3-(3-Trifluoromethylphenyl)propionic acid
CAS Number: 585-50-2
MDL Number: MFCD00090487
Molecular Formula: C10H9F3O2
Molecular Weight: 218.18
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-(Trifluoromethyl)hydrocinnamic Acid
Formula:
C₁₀H₉F₃O₂ MW: 218.18 g/mol Boiling Pt: 149 °C Melting Pt: 35 °C |
MDL Number:
MFCD00090487 CAS Number: 585-50-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...