Tris(4-fluorophenyl)phosphine ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
Tris(p-fluorophenyl)phosphine
T2900-5G
TCT2900-5GEA
145.15
USD
TCT2900-5G
Tris(4-fluorophenyl)phosphine ≥98.0% (by GC)
Tris(4-fluorophenyl)phosphine
CAS Number: 18437-78-0
MDL Number: MFCD00013553
Molecular Formula: C18H12F3P
Molecular Weight: 316.26
Purity/Analysis Method:
Form: Crystal
Color: White
Boiling point (°C): 160
Melting point (°C): 81
Formula:
C₁₈H₁₂F₃P MW: 316.26 g/mol Boiling Pt: 160 °C Melting Pt: 81 °C |
MDL Number:
MFCD00013553 CAS Number: 18437-78-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...