Thiourea ≥99.0% (by titrimetric analysis) for biochemical research
Supplier: TCI America
Synonyms:
Sulfourea, Thiocarbamide, Sulphourea
T2835-25G
T2835-5G
TCT2835-25GEA
47.01
USD
TCT2835-25G
TCT2835-5G
Thiourea ≥99.0% (by titrimetric analysis) for biochemical research
Thiourea
[for Biochemical Research]
CAS Number: 62-56-6
MDL Number: MFCD00008067
Molecular Formula: CH4N2S
Molecular Weight: 76.12
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Color: White
Melting point (°C): 177
Formula:
NH₂CSNH₂ MW: 76.12 g/mol Melting Pt: 177 °C |
MDL Number:
MFCD00008067 CAS Number: 62-56-6 UN: 3077 ADR: 9 III Merck Index: 13,09443 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...