Truxene ≥98.0%
Supplier: TCI America
Synonyms:
10,15-Dihydro-5H-tribenzo[a,f,k]trindene, Tribenzylenebenzene, 10,15-Dihydro-5H-diindeno[1,2-a:1',2'-c]fluorene
CAS Number: 548-35-6
MDL Number: MFCD00021180
Molecular Formula: C27H18
Molecular Weight: 342.44
Purity/Analysis Method:
Product Categories: Materials Chemistry, Nanocarbon Unit Structures (Nanocarbon Materials)
Other Product Names / Synonyms: Tribenzylenebenzene, 10,15-Dihydro-5H-tribenzo[a,f,k]trindene, 10,15-Dihydro-5H-diindeno[1,2-a:1',2'-c]fluorene
Formula:
C₂₇H₁₈ MW: 342.44 g/mol Melting Pt: 370 °C |
MDL Number:
MFCD00021180 CAS Number: 548-35-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...