Trioxsalen ≥98.0%
Supplier: TCI America
Synonyms:
4,5',8-Trimethylpsoralen, Trisoralen, TMP, 2,5,9-Trimethylfuro[3,2-g]benzopyran-7-one
CAS Number: 3902-71-4
Molecular Formula: C14H12O3
Molecular Weight: 228.25
Purity/Analysis Method:
Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Other Pharmaceutical Ingredients, Phenylpropanoids, Phenylpropanoids (Phytochemicals)
Other Product Names / Synonyms: TMP, 2,5,9-Trimethylfuro[3,2-g]benzopyran-7-one, 4,5',8-Trimethylpsoralen, Trisoralen
Formula:
C₁₄H₁₂O₃ MW: 228.25 g/mol Melting Pt: 234 °C |
MDL Number:
MFCD00005010 CAS Number: 3902-71-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...