6-Thioguanine ≥95.0% (by HPLC)
Supplier: TCI America
CAS Number: 154-42-7
MDL Number: MFCD00233553
Molecular Formula: C5H5N5S
Molecular Weight: 167.19
Purity/Analysis Method:
Product Categories: Bioscience, Immunosuppressant Ingredients, Antitumor Ingredients, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleobases and Their Analogs
Other Product Names / Synonyms: 2-Amino-6-mercaptopurine, 6-Mercaptoguanine
Formula:
C₅H₅N₅S |
MDL Number:
MFCD00233553 CAS Number: 154-42-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...