Streptomycin sulfate ≥95% (by total nitrogen and titration analysis)

Supplier: TCI America

S0834-25G S0834-5G
TCS0834-25GEA 36.54 USD
TCS0834-25G TCS0834-5G
Streptomycin sulfate ≥95% (by total nitrogen and titration analysis)
Streptomycin sulphate

Streptomycin sulfate is the first aminoglycoside antibiotic derived from the soil microbe Streptomyces griseus by Waksman and Schatz


  • Loss on drying: max. 5.0%


Streptomycin sulfate binds to the 30S subunit of the bacterial ribosome, and then it inhibits bacterial growth by inhibiting protein synthesis. Streptomycin-resistant bacterial strains appeared soon after its introduction.


Streptomycin sulfate has activity against aerobic Gram-negative bacteria that are not sensitive to the penicillins.


White crystals

Formula: C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
MW: 1457.4 g/mol
MDL Number: MFCD00037023
CAS Number: 3810-74-0
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=585b47fbc4a678504b2e6aea61e2844cf0bbfb8b&vid=cc56db0fd061629b15393267c021f729b28768d9&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR