Sodium triacetoxyborohydride ≥80.0%

Supplier: TCI America

S0394-100G S0394-25G
TCS0394-100GEA 203.99 USD
TCS0394-100G TCS0394-25G
Sodium triacetoxyborohydride ≥80.0%
Sodium triacetoxyborohydride

CAS Number: 56553-60-7 MDL Number: MFCD00012211 Molecular Formula: C6H11BO6 Molecular Weight: 211.94 Purity/Analysis Method: 80.0% (T) Form: Crystal Color: White Melting point (°C): 106

Formula: C₆H₁₀BNaO₆
Melting Pt: 106 °C
MDL Number: MFCD00012211
CAS Number: 56553-60-7
UN: 3396
ADR: 4.3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5674fe7b957ee8f986212c93b778865e48e2bec&vid=f5674fe7b957ee8f986212c93b778865e48e2bec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 80.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR