Quinidine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
(+)-Quinidine
CAS Number: 56-54-2
MDL Number: MFCD00135581
Molecular Formula: C20H24N2O2
Molecular Weight: 324.42
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Optical Resolving Agents, Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Alkaloids (Phytochemicals), Quinoline Alkaloids
Other Product Names / Synonyms: Conquinine
Formula:
C20H24N2O2 MW: 324.42 g/mol Melting Pt: 172 °C |
MDL Number:
MFCD00135581 CAS Number: 56-54-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...