Pentoxifylline ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
3,7-Dimethyl-1-(5-oxohexyl)xanthine, 1-(5-Oxohexyl)theobromine, 3,7-Dimethyl-1-(5-oxohexyl)-1H-purine-2,6(3H,7H)-dione, 3,7-Dihydro-3,7-dimethyl-1-(5-oxohexyl)-1H-purine-2,6-dione
CAS Number: 6493-05-6
MDL Number: MFCD00063379
Molecular Formula: C13H18N4O3
Molecular Weight: 278.31
Purity/Analysis Method:
Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 3,7-Dimethyl-1-(5-oxohexyl)xanthine, 3,7-Dihydro-3,7-dimethyl-1-(5-oxohexyl)-1H-purine-2,6-dione, 1-(5-Oxohexyl)theobromine
Formula:
C₁₃H₁₈N₄O₃ MW: 278.31 g/mol Melting Pt: 105 °C |
MDL Number:
MFCD00063379 CAS Number: 6493-05-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...