Piceatannol ≥98.0%

Supplier: TCI America

Synonyms: (E)-4-(3,5-Dihydroxystyryl)benzene-1,2-diol

P1928-100MG P1928-1G
TCP1928-100MGEA 223.94 USD
TCP1928-100MG TCP1928-1G
Piceatannol ≥98.0%
Piceatannol

CAS Number: 10083-24-6 MDL Number: MFCD00221715 Molecular Formula: C14H12O4 Molecular Weight: 244.25 Purity/Analysis Method: 98.0% (HPLC) Form: Crystal Color: White Boiling point (°C): 108 Melting point (°C): 235 Lambda max.: 323 nm (MeOH)Storage Temperature: 0-10°C


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Antioxidants, Aromatic Polyketides, Polyphenols (Phytochemicals), Histone Deacetylase Activators Other Product Names / Synonyms: 3,3',4,5'-Tetrahydroxy-trans-stilbene

Formula: C₁₄H₁₂O₄
Boiling Pt: 108 °C
Melting Pt: 235 °C
MDL Number: MFCD00221715
CAS Number: 10083-24-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85d032f03f8ce5cd6a2f49cd44a2927f07f3224e&vid=85d032f03f8ce5cd6a2f49cd44a2927f07f3224e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR