Polymyxin B sulphate

Supplier: TCI America

P1923-1G P1923-5G
TCP1923-1GEA 150.16 USD
TCP1923-1G TCP1923-5G
Polymyxin B sulphate
Polymyxin B sulphate

Polymyxin B sulfate is a polypeptide antibiotic that targets gram negative bacteria.


  • White crystals

Formula: C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
MW: 1385.61 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00131991
CAS Number: 1405-20-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76b8914a5dcfb7951ce8c1eecca44acf9058ab9a&vid=76b8914a5dcfb7951ce8c1eecca44acf9058ab9a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR