1,2,2,3-Propanetetracarbonitrile ≥96.0% (by GC)

Supplier: TCI America

Synonyms: 1,2,2,3-Tetracyanopropane

P1751-1G P1751-5G
TCP1751-1GEA 207 USD
TCP1751-1G TCP1751-5G
1,2,2,3-Propanetetracarbonitrile ≥96.0% (by GC)
1,2,2,3-Propanetetracarbonitrile

CAS Number: 1274904-48-1 Molecular Formula: C7H4N4 Molecular Weight: 144.14 Purity/Analysis Method: 96.0% (GC) Form: Crystal Color: Very Pale Yellow


Product Categories: Materials Chemistry, Secondary Battery Research Reagents Other Product Names / Synonyms: 1,2,2,3-Tetracyanopropane

Formula: C₇H₄N₄
MW: 144.14 g/mol
CAS Number: 1274904-48-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70940d05808ce007ec12f41d329731715ffdd2c2&vid=70940d05808ce007ec12f41d329731715ffdd2c2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR