Pentafluorobenzene ≥98.0%
Supplier: TCI America
P0923-25G
P0923-5G
TCP0923-25GEA
82
USD
TCP0923-25G
TCP0923-5G
Pentafluorobenzene ≥98.0%
Pentafluorobenzene
CAS Number: 363-72-4
MDL Number: MFCD00000286
Molecular Formula: C6HF5
Molecular Weight: 168.07
Purity/Analysis Method:
Formula:
C₆HF₅ Boiling Pt: 85 °C Melting Pt: –48 °C Density: 1.53 (20 °C) Flash Pt: 13 °C |
MDL Number:
MFCD00000286 CAS Number: 363-72-4 UN: 1993 ADR: 3,II |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...