Pilocarpine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
(+)-Pilocarpine hydrochloride
P0434-1G
P0434-5G
TCP0434-1GEA
101.56
USD
TCP0434-1G
TCP0434-5G
Pilocarpine hydrochloride ≥98.0% (by HPLC, titration analysis)
Pilocarpine hydrochloride
CAS Number: 54-71-7
MDL Number: MFCD00012722
Molecular Formula: C11H16N2O2
Molecular Weight: 244.72
Purity/Analysis Method:
Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Alkaloids (Phytochemicals), Imidazole Alkaloids
Other Product Names / Synonyms: 3-Ethyl-4-[(1-methyl-5-imidazolyl)methyl]tetrahydrofuran-2-one Hydrochloride
Formula:
C₁₁H₁₇ClN₂O₂ Melting Pt: 203 °C |
MDL Number:
MFCD00012722 CAS Number: 54-71-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...