Phenylhydrazinium chloride ≥98.5%

Supplier: TCI America

Synonyms: Phenylhydrazine hydrochloride, Phenylhydrazinium chloride

P0184-25G P0184-100G
TCP0184-25GEA 34.91 USD
TCP0184-25G TCP0184-100G
Phenylhydrazinium chloride ≥98.5%
Phenylhydrazinium chloride

CAS Number: 59-88-1 MDL Number: MFCD00012924 Molecular Formula: C6H8N2 Molecular Weight: 144.60 Purity/Analysis Method: 98.5% (T) Form: Crystal Melting point (°C): 254


Other Product Names / Synonyms: Phenylhydrazinium Chloride

Formula: C₆H₉ClN₂
MW: 144.6 g/mol
Melting Pt: 254 °C
MDL Number: MFCD00012924
CAS Number: 59-88-1
UN: 2811
ADR: 6.1 III
Merck Index: 12,07447
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87cf1828fd97609f2f1d0bbaeb0d9c2c10a3ff27&vid=87cf1828fd97609f2f1d0bbaeb0d9c2c10a3ff27&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.5% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR