1H,1H,5H-Octafluoropentyl 1,1,2,2-tetrafluoroethyl ether ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
2,2,3,3,4,4,5,5-Octafluoropentyl 1,1,2,2-Tetrafluoroethyl Ether, 1,1,2,2,3,3,4,4-Octafluoro-5-(1,1,2,2-tetrafluoroethoxy)pentane
CAS Number: 16627-71-7
MDL Number: MFCD00155904
Molecular Formula: C7H4F12O
Molecular Weight: 332.09
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: 2,2,3,3,4,4,5,5-Octafluoropentyl 1,1,2,2-Tetrafluoroethyl Ether
Formula:
C₇H₄F₁₂O MW: 332.09 g/mol Boiling Pt: 133 °C Density: 1.66 (20 °C) |
MDL Number:
MFCD00155904 CAS Number: 16627-71-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...