Nebivolol hydrochloride ≥98.0%
Supplier: TCI America
N0954-100MG
N0954-20MG
TCN0954-100MGEA
213
USD
TCN0954-100MG
TCN0954-20MG
Nebivolol hydrochloride ≥98.0%
Nebivolol hydrochloride
CAS Number: 152520-56-4
MDL Number: MFCD07357273
Molecular Formula: C22H25F2NO4
Molecular Weight: 441.90
Purity/Analysis Method:
Formula:
C₂₂H₂₅F₂NO₄ Melting Pt: 226 °C |
MDL Number:
MFCD07357273 CAS Number: 152520-56-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...