4-Methyl-1,3,2-dioxathiolane-2-oxide (mixture of isomers) ≥98.0% (by GC)

Supplier: TCI America

M2471-25G M2471-5G
TCM2471-25GEA 137 USD
TCM2471-25G TCM2471-5G
4-Methyl-1,3,2-dioxathiolane-2-oxide (mixture of isomers) ≥98.0% (by GC)
4-Methyl-1,3,2-dioxathiolane-2-oxide (mixture of isomers)

CAS Number: 1469-73-4
MDL Number: MFCD00041209
Molecular Formula: C3H6O3S
Molecular Weight: 122.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 40
Flash Point (°C): 81
Specific Gravity (20/20): 1.30


Product Categories: Synthetic Organic Chemistry, Others (Sulfur Compounds for Synthesis), Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: 1,2-Propanediol Cyclic Sulfite, 1,2-Propyleneglycol Sulfite, 1,2-Propylene Sulfite

Formula: C₃H₆O₃S
Boiling Pt: 40 °C
Density: 1.30 (20 °C)
Flash Pt: 81 °C
MDL Number: MFCD00041209
CAS Number: 1469-73-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b6ac577e14a711702b19c10d18e90c9634c1e14&vid=5b6ac577e14a711702b19c10d18e90c9634c1e14&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR