Mizoribine ≥98.0%
Supplier: TCI America
Synonyms:
1-(beta-D-Ribofuranosyl)-5-hydroxyimidazole-4-carboxamide, 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-hydroxy-1H-imidazole-4-carboxamide
CAS Number: 50924-49-7
MDL Number: MFCD00057221
Molecular Formula: C9H13N3O6
Molecular Weight: 259.22
Purity/Analysis Method:
Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Immunosuppressant Ingredients, Anti-rheumatoid Arthritis Ingredients, Antiviral Ingredients, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleo
Other Product Names / Synonyms: 1-(beta-D-Ribofuranosyl)-5-hydroxyimidazole-4-carboxamide
Formula:
C₉H₁₃N₃O₆ MW: 259.22 g/mol Melting Pt: 198 °C |
MDL Number:
MFCD00057221 CAS Number: 50924-49-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...