4-Methoxy-2-methylphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
CAS Number: 208399-66-0
MDL Number: MFCD02684315
Molecular Formula: C8H11BO3
Molecular Weight: 165.98
Form: Crystal
Color: White
Melting point (°C): 191
Other Product Names / Synonyms: 4-Methoxy-2-methylbenzeneboronic Acid
Formula:
C₈H₁₁BO₃ Melting Pt: 191 °C |
MDL Number:
MFCD02684315 CAS Number: 208399-66-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...