N-Methylsuccinimide ≥98.0% (by GC)
Supplier: TCI America
M2207-25G
M2207-5G
TCM2207-25GEA
587.76
USD
TCM2207-25G
TCM2207-5G
N-Methylsuccinimide ≥98.0% (by GC)
N-Methylsuccinimide
CAS Number: 1121-07-9
MDL Number: MFCD00005517
Molecular Formula: C5H7NO2
Molecular Weight: 113.12
Purity/Analysis Method:
Formula:
C₅H₇NO₂ Boiling Pt: 235 °C Melting Pt: 70 °C |
MDL Number:
MFCD00005517 CAS Number: 1121-07-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...