3-Methyl-2-benzothiazolinonehydrazone hydrochloride hydrate ≥98.0% (by HPLC, titration analysis) for biochemical research

Supplier: TCI America

M2155-1G M2155-5G
TCM2155-1GEA 19 USD
TCM2155-1G TCM2155-5G
3-Methyl-2-benzothiazolinonehydrazone hydrochloride hydrate ≥98.0% (by HPLC, titration analysis) for biochemical research
3-Methyl-2-benzothiazolinonehydrazone hydrochloride hydrate

[for Biochemical Research]
CAS Number: 149022-15-1
MDL Number: MFCD00149370
Molecular Formula: C8H9N3S
Molecular Weight: 215.70
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Product Categories: Bioscience, Peroxidase Substrates, Redox Research Reagents
Other Product Names / Synonyms: MBTH Hydrochloride, Sawicki's Reagent

Formula: C₈H₉N₃S
MDL Number: MFCD00149370
CAS Number: 149022-15-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e073b45c96840b3f5fd34c99618b7153873c562&vid=f7e2399174dd41c66701c45e48d62931a25f9386&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR