2-Methoxy-5-pyridineboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
CAS Number: 163105-89-3
MDL Number: MFCD02093044
Molecular Formula: C6H8BNO3
Molecular Weight: 152.94
Form: Crystal
Color: White
Melting point (°C): 138
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 6-Methoxy-3-pyridylboronic Acid
Formula:
C₆H₈BNO₃ Melting Pt: 138 °C |
MDL Number:
MFCD02093044 CAS Number: 163105-89-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...