Cyclopentyl methyl ether (CPME) ≥99.5% (by GC) stabilized

Supplier: TCI America

M1698-100ML M1698-500ML
TCM1698-100MLEA 21.69 USD
TCM1698-100ML TCM1698-500ML
Cyclopentyl methyl ether (CPME) ≥99.5% (by GC) stabilized
Cyclopentyl methyl ether (CPME)

(stabilized with BHT) CAS Number: 5614-37-9 Molecular Formula: C6H12O Molecular Weight: 100.16 Purity/Analysis Method: 99.5% (GC) Form: Clear Liquid Boiling point (°C): 106 Melting point (°C): -134 Flash Point (°C): -1 Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: Cyclopentyl Methyl Ether, CPME

Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 106 °C
Melting Pt: –134 °C
Density: 0.86 (20 °C)
Flash Pt: –1 °C
MDL Number: MFCD08276401
CAS Number: 5614-37-9
UN: 3271
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7ebc52e88bcb888e42d16fa603751eef38c4e1d&vid=b7ebc52e88bcb888e42d16fa603751eef38c4e1d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR