(±)-2-Methyltetrahydrofuran-3-one ≥98.0% (by GC)

Supplier: TCI America

M1055-25G M1055-5G
TCM1055-25GEA 93 USD
TCM1055-25G TCM1055-5G
(±)-2-Methyltetrahydrofuran-3-one ≥98.0% (by GC)
(±)-2-Methyltetrahydrofuran-3-one

CAS Number: 3188-00-9 MDL Number: MFCD00010423 Molecular Formula: C5H8O2 Molecular Weight: 100.12 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 139 Flash Point (°C): 38 Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: Tetrahydro-2-methylfuran-3-one

Formula: C₅H₈O₂
Boiling Pt: 139 °C
Density: 1.04 (20 °C)
Flash Pt: 38 °C
MDL Number: MFCD00010423
CAS Number: 3188-00-9
UN: 1224
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1530903e8e230acae03334ba5daf567f8c1e17a&vid=b1530903e8e230acae03334ba5daf567f8c1e17a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR