L-tert-Leucine methyl ester ≥98.0%
Supplier: TCI America
Synonyms:
(S)-Methyl 2-amino-3,3-dimethylbutanoate, (S)-2-Amino-3,3-dimethylbutyric Acid Methyl Ester, H-Tle-OMe, Methyl (S)-2-Amino-3,3-dimethylbutyrate
CAS Number: 63038-26-6
MDL Number: MFCD00137409
Molecular Formula: C7H15NO2
Molecular Weight: 145.20
Purity/Analysis Method:
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Tle-OMe, Methyl (S)-2-Amino-3,3-dimethylbutyrate, (S)-2-Amino-3,3-dimethylbutyric Acid Methyl Ester
Formula:
C₇H₁₅NO₂ Boiling Pt: 69 °C |
MDL Number:
MFCD00137409 CAS Number: 63038-26-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...