Hydrogen chloride ∼1 mol/L in ethyl acetate

Supplier: TCI America

H1060-25ML H1060-500ML
TCH1060-25MLEA 31.05 USD
TCH1060-25ML TCH1060-500ML
Hydrogen chloride ∼1 mol/L in ethyl acetate
Hydrogen chloride

CAS Number: 7647-01-0
MDL Number: MFCD00011324
Molecular Formula: HCl
Molecular Weight: 36.46
Form: Clear Liquid
Color: Colorless
Storage Temperature: 0-10°C

Formula: HCl
MW: 36.46
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1050
ADR: 2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR