Guanidinium nitrate ≥98.0% (by gravimetric analysis)

Supplier: TCI America

Synonyms: Guanidine nitrate

G0164-25G
TCG0164-25GEA 19.87 USD
TCG0164-25G
Guanidinium nitrate ≥98.0% (by gravimetric analysis)
Guanidinium nitrate

CAS Number: 506-93-4
MDL Number: MFCD00013028
Molecular Formula: CH5N3
Molecular Weight: 122.08
Purity/Analysis Method: 98.0% (W)
Form: Crystal
Melting point (°C): 214

Formula: CH₆N₄O₃
MW: 122.08 g/mol
Melting Pt: 214 °C
MDL Number: MFCD00013028
CAS Number: 506-93-4
UN: 1467
ADR: 5.1,III
https://us-prod2.vwr.com/stibo/web/std.lang.all/98/22/28969822.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (W)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR