Fenofibrate ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 49562-28-9
MDL Number: MFCD00133314
Molecular Formula: C20H21ClO4
Molecular Weight: 360.83
Purity/Analysis Method:
Product Categories: Bioscience, Antilipemic Ingredients
Other Product Names / Synonyms: 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic Acid Isopropyl Ester, Isopropyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionate
Formula:
C₂₀H₂₁ClO₄ Melting Pt: 81 °C |
MDL Number:
MFCD00133314 CAS Number: 49562-28-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...