(±)-Ethyl-2-bromovalerate 98,0

Supplier: TCI America

Synonyms: Ethyl 2-bromovalerate

E0970-100G E0970-25G
TCE0970-100GEA 143.3 USD
TCE0970-100G TCE0970-25G
(±)-Ethyl-2-bromovalerate 98,0
(±)-Ethyl 2-bromovalerate

CAS Number: 615-83-8 MDL Number: MFCD00000159 Molecular Formula: C7H13BrO2 Molecular Weight: 209.08 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 94 Flash Point (°C): 77 Specific Gravity (20/20): 1.28


Other Product Names / Synonyms: 2-Bromovaleric Acid Ethyl Ester

Formula: C₇H₁₃BrO₂
MW: 209.09 g/mol
Boiling Pt: 94 °C
Density: 1.28 (20 °C)
Flash Pt: 77 °C
MDL Number: MFCD00000159
CAS Number: 615-83-8
UN: 3265
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ff568babda28582a81db0e7a998955744bdb370&vid=5ff568babda28582a81db0e7a998955744bdb370&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR