1,2-Epoxy-4-vinylcyclohexane (mixture of isomers) ≥98.0%

Supplier: TCI America

Synonyms: 3-Vinyl-7-oxabicyclo[4.1.0]heptane, 4-Vinyl-1,2-epoxycyclohexane

E0956-250ML E0956-25ML
TCE0956-250MLEA 102.6 USD
TCE0956-250ML TCE0956-25ML
1,2-Epoxy-4-vinylcyclohexane (mixture of isomers) ≥98.0%
1,2-Epoxy-4-vinylcyclohexane (mixture of isomers)

CAS Number: 106-86-5 MDL Number: MFCD00022356 Molecular Formula: C8H12O Molecular Weight: 124.18 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 159 Melting point (°C): -100 Flash Point (°C): 58 Specific Gravity (20/20): 0.96 Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers Other Product Names / Synonyms: 4-Vinyl-1,2-epoxycyclohexane, 3-Vinyl-7-oxabicyclo[4.1.0]heptane

Formula: C₈H₁₂O
Boiling Pt: 159 °C
Melting Pt: –100 °C
Density: 0.96 (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00022356
CAS Number: 106-86-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f075bf2cc484d06313165f5eadb0f5ad20eba8b&vid=7f075bf2cc484d06313165f5eadb0f5ad20eba8b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR