Ebselen ≥98.0%

Supplier: TCI America

Synonyms: 2-Phenyl-1,2-benzoisoselenazol-3(2H)-one, 2-Phenylbenzo[d][1,2]selenazol-3(2H)-one

E0946-100MG E0946-25MG
TCE0946-100MGEA 320.92 USD
TCE0946-100MG TCE0946-25MG
Ebselen ≥98.0%
Ebselen

CAS Number: 60940-34-3 MDL Number: MFCD00210937 Molecular Formula: C13H9NOSe Molecular Weight: 274.18 Purity/Analysis Method: 98.0% (GC) Form: Crystal Melting point (°C): 183 Flash Point (°C): 197 Lambda max.: 327 nm (MeOH)Storage Temperature: 0-10°C


Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Antioxidants, ROS/RNS-related Enzyme Inhibitors, Other Pharmaceutical Ingredients Other Product Names / Synonyms: 2-Phenyl-1,2-benzoisoselenazol-3(2H)-one

Formula: C₁₃H₉NOSe
MW: 274.20 g/mol
Melting Pt: 183 °C
Flash Pt: 197 °C
MDL Number: MFCD00210937
CAS Number: 60940-34-3
UN: 3283
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c34fa57cc9e8d31451bac50b80311de0276a56c3&vid=c34fa57cc9e8d31451bac50b80311de0276a56c3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR