N-Ethylpropylamine ≥97.0% (by GC)

Supplier: TCI America

E0351-250ML E0351-25ML
TCE0351-250MLEA 1154 USD
TCE0351-250ML TCE0351-25ML
N-Ethylpropylamine ≥97.0% (by GC)
N-Ethylpropylamine

CAS Number: 20193-20-8 MDL Number: MFCD00015198 Molecular Formula: C5H13N Molecular Weight: 87.17 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Boiling point (°C): 79 Flash Point (°C): 4 Specific Gravity (20/20): 0.72

Formula: C₅H₁₃N
Boiling Pt: 79 °C
Density: 0.72 (20 °C)
Flash Pt: 4 °C
MDL Number: MFCD00015198
CAS Number: 20193-20-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea82e8f96a78a7e503d58df1edea0d8afe038efa&vid=ea82e8f96a78a7e503d58df1edea0d8afe038efa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR