3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)] ≥98.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
DiSC3(5), 3-Propyl-2-[5-[3-propyl-2(3H)-benzothiazolylidene]-1,3-pentadienyl]benzothiazolium iodide, 5-(3-Propylbenzothiazol-2-ylidene)-1-(3-propylbenzothiazolium-2-yl)-1,3-pentadiene odide, 3,3'-Dipropylthiadicarbocyanine iodide
CAS Number: 53213-94-8
MDL Number: MFCD00075033
Molecular Formula: C25H27IN2S2
Molecular Weight: 546.53
Purity/Analysis Method:
Product Categories: Materials Chemistry, Cyanine & Squarylium Dyes (Functional Organic Dyes), Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: DiSC3(5), 3-Propyl-2-[5-[3-propyl-2(3H)-benzothiazolylidene]-1,3-pentadienyl]benzothiazolium Iodide, 5-(3-Propylbenzothiazol-2-ylidene)-1-(3-propylbenzothiazolium-2-yl)-1,3-pentadiene Iodide
Formula:
C₂₅H₂₇IN₂S₂ MW: 546.53 g/mol Melting Pt: 245 °C |
MDL Number:
MFCD00075033 CAS Number: 53213-94-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...