N,N-Diethylformamide ≥99.0% (by GC) for biochemical research

Supplier: TCI America

Synonyms: Formic acid diethylamide, Formdiethylamide

D4234-500ML D4234-100ML
TCD4234-500MLEA 383 USD
TCD4234-500ML TCD4234-100ML
N,N-Diethylformamide ≥99.0% (by GC) for biochemical research
N,N-Diethylformamide

[for Biochemical Research]
CAS Number: 617-84-5
MDL Number: MFCD00003287
Molecular Formula: C5H11NO
Molecular Weight: 101.15
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 177
Flash Point (°C): 60
Specific Gravity (20/20): 0.91


Other Product Names / Synonyms: Formyldiethylamine

Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 176
Density: 0.91 (20 °C)
Flash Pt: 60 °C
MDL Number: MFCD00003287
CAS Number: 617-84-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0be30b8c35a9bb256049994de2cbbd6d61325c47&vid=ded8de3b5455b03544a0cbe497007667d0a1c92c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR