Daphnetin ≥90.0% (by HPLC)
Supplier: TCI America
Synonyms:
7,8-Dihydroxy-2H-chromen-2-one, 7,8-Dihydroxycoumarin
D4001-1G
D4001-5G
TCD4001-1GEA
177.8
USD
TCD4001-1G
TCD4001-5G
Daphnetin ≥90.0% (by HPLC)
Daphnetin
CAS Number: 486-35-1
MDL Number: MFCD00016977
Molecular Formula: C9H6O4
Molecular Weight: 178.14
Purity/Analysis Method:
Product Categories: Bioscience, Phenylpropanoids, Phenylpropanoids (Phytochemicals)
Other Product Names / Synonyms: 7,8-Dihydroxycoumarin
Formula:
C₉H₆O₄ MW: 178.14 g/mol Melting Pt: 256 °C |
MDL Number:
MFCD00016977 CAS Number: 486-35-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...