Diosmin ≥85.0%
Supplier: TCI America
Synonyms:
5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one
CAS Number: 520-27-4
MDL Number: MFCD00009772
Molecular Formula: C28H32O15
Molecular Weight: 608.55
Purity/Analysis Method:
Product Categories: Bioscience, Glycosides, Disaccharides, Glycosides, Antioxidants, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: Barosmin, 3',5,7-Trihydroxy-4'-methoxyflavone 7-Rutinoside
Formula:
C₂₈H₃₂O₁₅ MW: 608.5 g/mol Melting Pt: 280 °C |
MDL Number:
MFCD00009772 CAS Number: 520-27-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...