Borane dimethyl sulfide complex ≥90.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Borane-methyl sulfide complex, Borane-methyl sulphide complex
D1843-25ML
D1843-100ML
TCD1843-25MLEA
47.05
USD
TCD1843-25ML
TCD1843-100ML
Borane dimethyl sulfide complex ≥90.0% (by titrimetric analysis)
Borane dimethyl sulphide complex
CAS Number: 13292-87-0
MDL Number: MFCD00013189
Molecular Formula: C2H9BS
Molecular Weight: 75.96
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Reduction
Other Product Names / Synonyms: Borane - Dimethyl Sulfide Complex, BMS, Methyl Sulfide Borane
Formula:
C₂H₉BS MW: 75.97 g/mol Density: 0.80 (20 °C) Flash Pt: 18 °C |
MDL Number:
MFCD00013189 CAS Number: 13292-87-0 UN: 2924 ADR: 3,II |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...