1,3-Dichloropropane
Supplier: TCI America
Synonyms:
Trimethylendichloride
D0399-250ML
D0399-25ML
TCD0399-250MLEA
319.62
USD
TCD0399-250ML
TCD0399-25ML
1,3-Dichloropropane
1,3-Dichloropropane
CAS Number: 142-28-9
MDL Number: MFCD00000999
Molecular Formula: C3H6Cl2
Molecular Weight: 112.98
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 122
Flash Point (°C): 21
Specific Gravity (20/20): 1.19
Other Product Names / Synonyms: Trimethylene Chloride
Formula:
ClH₂CCH₂CH₂Cl MW: 112.99 Boiling Pt: 120.4 Melting Pt: -99.5 Density: 1.19 (20 °C) Flash Pt: 21 °C |
MDL Number:
MFCD00000999 CAS Number: 142-28-9 UN: 1993 ADR: 3 II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...