Corannulene ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
Dibenzo[ghi,mno]fluoranthene
C2572-100MG
C2572-20MG
TCC2572-100MGEA
1026.3
USD
TCC2572-100MG
TCC2572-20MG
Corannulene ≥97.0% (by GC)
Corannulene
CAS Number: 5821-51-2
Molecular Formula: C20H10
Molecular Weight: 250.30
Purity/Analysis Method:
Product Categories: Materials Chemistry, Nanocarbon Unit Structures (Nanocarbon Materials)
Other Product Names / Synonyms: Dibenzo[ghi,mno]fluoranthene
Formula:
C₂₀H₁₀ MW: 250.30 g/mol Melting Pt: 269 °C |
MDL Number:
MFCD22124717 CAS Number: 5821-51-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...