Clofarabine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
2-Chloro-2'-arabino-fluoro-2'-deoxyadenosine, (2R,3R,4S,5R)-5-(6-Amino-2-chloro-9H-purin-9-yl)-4-fluoro-2-(hydroxymethyl)tetrahydrofuran-3-ol, 2-Chloro-9-(2-deoxy-2-fluoro-beta-D-arabinofuranosyl)adenine
CAS Number: 123318-82-1
MDL Number: MFCD00871077
Molecular Formula: C10H11ClFN5O3
Molecular Weight: 303.68
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Antitumor Ingredients, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleosides and Their Analogs
Other Product Names / Synonyms: 2-Chloro-9-(2-deoxy-2-fluoro-beta-D-arabinofuranosyl)adenine
Formula:
C₁₀H₁₁ClFN₅O₃ MW: 303.68 g/mol Melting Pt: 227 °C |
MDL Number:
MFCD00871077 CAS Number: 123318-82-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...