Isoflurane ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether

C2485-25G C2485-5G
TCC2485-25GEA 192.36 USD
TCC2485-25G TCC2485-5G
Isoflurane ≥98.0% (by GC)
Isoflurane

CAS Number: 26675-46-7 MDL Number: MFCD00066609 Molecular Formula: C3H2ClF5O Molecular Weight: 184.49 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 49 Specific Gravity (20/20): 1.51


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Bioactive Compounds, Fluorinated Building Blocks, Bioscience, Other Pharmaceutical Ingredients Other Product Names / Synonyms: 2-Chloro-2-(difluoromethoxy)-1,1,1-trifluoroethane, Isoflurane

Formula: C₃H₂ClF₅O
MW: 184.49 g/mol
Boiling Pt: 49 °C
Density: 1.51 (20 °C)
MDL Number: MFCD00066609
CAS Number: 26675-46-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b96833421ef1a4ec4f5bd47b8cdc4296e37bb97&vid=3b96833421ef1a4ec4f5bd47b8cdc4296e37bb97&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR