5-Chloronicotinic acid ≥95.0%
Supplier: TCI America
C2399-1G
C2399-5G
TCC2399-1GEA
38
USD
TCC2399-1G
TCC2399-5G
5-Chloronicotinic acid ≥95.0%
5-Chloronicotinic acid
CAS Number: 22620-27-5
MDL Number: MFCD00128901
Molecular Formula: C6H4ClNO2
Molecular Weight: 157.55
Purity/Analysis Method:
Other Product Names / Synonyms: 5-Chloro-3-pyridinecarboxylic Acid
Formula:
C₆H₄ClNO₂ MW: 157.56 Melting Pt: 171 °C |
MDL Number:
MFCD00128901 CAS Number: 22620-27-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...