Carbonylchlorobis(triphenylphosphine)iridium(I) ≥95.0%
Supplier: TCI America
Synonyms:
Carbonylchloro bis(triphenylphosphine)iridium(I), VASKA'S COMPLEX, Chlorocarbonylbis(triphenylphosphine)iridium(I)
CAS Number: 14871-41-1
MDL Number: MFCD00009590
Molecular Formula: C37H30ClIrOP2
Molecular Weight: 780.26
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Activation Reaction of Unactivated Bonds
Other Product Names / Synonyms: Bis(triphenylphosphine)iridium(I) Carbonyl Chloride, Carbonylbis(triphenylphosphine)iridium(I) Chloride, Vaska's Catalyst
Formula:
C₃₇H₃₀ClIrOP₂ Melting Pt: 225 °C |
MDL Number:
MFCD00009590 CAS Number: 14871-41-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...